Assay Details
| Gene Symbol: HEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746927.4 | XP_002746973.1 | 4 - 5 | 540 | 70 | |
| XM_008994992.2 | XP_008993240.1 | 4 - 5 | 391 | 70 | ||
More Information
Set Membership: |
|

HEY2
Genomic Map