Assay Details
Gene Symbol: KDELC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002742523.3 | XP_002742569.1 | 2 - 3 | 774 | 67 | |
XM_017970378.1 | XP_017825867.1 | 2 - 3 | 445 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
