Assay Details
| Gene Symbol: CLPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761622.3 | XP_002761668.1 | 5 - 6 | 774 | 105 | |
| XM_017968036.1 | XP_017823525.1 | 4 - 5 | 633 | 105 | ||
More Information
Set Membership: |
|

CLPP
Genomic Map