Assay Details
| Gene Symbol: NELFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006057.2 | XP_009004305.2 | 10 - 11 | 1422 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NELFB| Gene Symbol: NELFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006057.2 | XP_009004305.2 | 10 - 11 | 1422 | 79 | |
Set Membership: |
|