Assay Details
| Gene Symbol: SS18L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735089.3 | XP_003735137.1 | 2 - 3 | 202 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SS18L2| Gene Symbol: SS18L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735089.3 | XP_003735137.1 | 2 - 3 | 202 | 103 | |
Set Membership: |
|