Assay Details
| Gene Symbol: ADAMTSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968119.1 | XP_017823608.1 | 8 - 9 | 1218 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADAMTSL5| Gene Symbol: ADAMTSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968119.1 | XP_017823608.1 | 8 - 9 | 1218 | 61 | |
Set Membership: |
|