Assay Details
| Gene Symbol: LOC100399567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749016.4 | XP_002749062.2 | 1 | 1655 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100399567| Gene Symbol: LOC100399567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749016.4 | XP_002749062.2 | 1 | 1655 | 99 | |
Set Membership: |
|