Assay Details
| Gene Symbol: GLYCTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758414.3 | XP_002758460.1 | 4 - 5 | 775 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLYCTK| Gene Symbol: GLYCTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758414.3 | XP_002758460.1 | 4 - 5 | 775 | 89 | |
Set Membership: |
|