Assay Details
| Gene Symbol: ZHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759254.4 | XP_002759300.1 | 3 - 4 | 3601 | 107 | |
| XM_003735014.3 | XP_003735062.1 | 3 - 4 | 3696 | 107 | ||
| XM_008983294.2 | XP_008981542.1 | 4 - 5 | 3656 | 107 | ||
More Information
Set Membership: |
|

ZHX1
Genomic Map