Assay Details
Gene Symbol: HAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762012.4 | XP_002762058.1 | 3 - 4 | 432 | 78 | |
GenBank mRNA | EU076445.1 | - | 2 - 3 | 188 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
