Assay Details
| Gene Symbol: MOB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742987.3 | XP_002743033.1 | 2 - 3 | 911 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOB3B| Gene Symbol: MOB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742987.3 | XP_002743033.1 | 2 - 3 | 911 | 92 | |
Set Membership: |
|