Assay Details
| Gene Symbol: HOMER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749176.4 | XP_002749222.1 | 2 - 3 | 323 | 65 | |
| XM_008998485.2 | XP_008996733.1 | 2 - 3 | 327 | 65 | ||
More Information
Set Membership: |
|

HOMER2
Genomic Map