Assay Details
Gene Symbol: SIKE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002751267.4 | XP_002751313.1 | 3 - 4 | 491 | 77 | |
XM_009001945.2 | XP_009000193.1 | 3 - 4 | 504 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
