Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755553.4 | XP_002755599.1 | 2 - 3 | 407 | 78 | |
| XM_009008506.2 | XP_009006754.1 | 2 - 3 | 380 | 78 | ||
More Information
Set Membership: |
|

GAL3ST3
Genomic Map