Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760935.4 | XP_002760981.3 | 7 - 8 | 1399 | 83 | |
| XM_002760936.3 | XP_002760982.1 | 7 - 8 | 1399 | 83 | ||
| XM_008985846.2 | XP_008984094.1 | 7 - 8 | 1399 | 83 | ||
More Information
Set Membership: |
|

TENT4B
Genomic Map