Assay Details
| Gene Symbol: ZSCAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970810.1 | XP_017826299.1 | 1 - 2 | 114 | 111 | |
| XM_017970811.1 | XP_017826300.1 | 1 - 2 | 114 | 111 | ||
| XM_017970812.1 | XP_017826301.1 | 1 - 2 | 114 | 111 | ||
| XM_017970813.1 | XP_017826302.1 | 1 - 2 | 114 | 111 | ||
| XM_017970814.1 | XP_017826303.1 | 1 - 2 | 114 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017970810.1, XM_017970811.1, XM_017970812.1, XM_017970813.1, XM_017970814.1 transcripts.
Set Membership: |
|

ZSCAN12
Genomic Map