Assay Details
| Gene Symbol: TANGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743550.3 | XP_002743596.1 | 7 - 8 | 719 | 90 | |
| XM_009006858.2 | XP_009005106.1 | 8 - 9 | 766 | 90 | ||
| XM_009006884.2 | XP_009005132.1 | 7 - 8 | 685 | 90 | ||
| XM_009006950.2 | XP_009005198.1 | 5 - 6 | 481 | 90 | ||
| XM_009006954.1 | XP_009005202.1 | 6 - 7 | 592 | 90 | ||
| XM_017977995.1 | XP_017833484.1 | 8 - 9 | 766 | 90 | ||
| XM_017977998.1 | XP_017833487.1 | 8 - 9 | 813 | 90 | ||
| XM_017978007.1 | XP_017833496.1 | 8 - 9 | 820 | 90 | ||
| XM_017978011.1 | XP_017833500.1 | 7 - 8 | 754 | 90 | ||
| XM_017978015.1 | XP_017833504.1 | 7 - 8 | 685 | 90 | ||
| XM_017978016.1 | XP_017833505.1 | 7 - 8 | 814 | 90 | ||
| XM_017978017.1 | XP_017833506.1 | 7 - 8 | 742 | 90 | ||
| XM_017978018.1 | XP_017833507.1 | 6 - 7 | 673 | 90 | ||
| XM_017978023.1 | XP_017833512.1 | 6 - 7 | 673 | 90 | ||
| XM_017978032.1 | XP_017833521.1 | 5 - 6 | 518 | 90 | ||
| XM_017978033.1 | XP_017833522.1 | 5 - 6 | 481 | 90 | ||
| XM_017978038.1 | XP_017833527.1 | 5 - 6 | 518 | 90 | ||
| XM_017978048.1 | XP_017833537.1 | 5 - 6 | 535 | 90 | ||
More Information
Set Membership: |
|

TANGO2
Genomic Map