Assay Details
| Gene Symbol: ZNF232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997264.2 | XP_008995512.1 | 3 - 4 | 857 | 87 | |
| XM_017972606.1 | XP_017828095.1 | 3 - 4 | 539 | 87 | ||
More Information
Set Membership: |
|

ZNF232
Genomic Map