Assay Details
| Gene Symbol: LOC100389034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994676.2 | XP_008992924.1 | 3 - 4 | 307 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100389034| Gene Symbol: LOC100389034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994676.2 | XP_008992924.1 | 3 - 4 | 307 | 96 | |
Set Membership: |
|