Assay Details
| Gene Symbol: TEDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754330.3 | XP_002754376.2 | 7 - 8 | 1491 | 95 | |
| XM_017977394.1 | XP_017832883.1 | 7 - 8 | 923 | 95 | ||
More Information
Set Membership: |
|

TEDC1
Genomic Map