Assay Details
Gene Symbol: LOC100391167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008983190.2 | XP_008981438.1 | 3 - 4 | 838 | 85 | |
XM_008983196.2 | XP_008981444.1 | 3 - 4 | 768 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
