Assay Details
| Gene Symbol: LOC100391167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983190.2 | XP_008981438.1 | 4 - 5 | 1048 | 66 | |
| XM_008983196.2 | XP_008981444.1 | 4 - 5 | 978 | 66 | ||
More Information
Set Membership: |
|

LOC100391167
Genomic Map