Assay Details
| Gene Symbol: LOC100391167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983190.2 | XP_008981438.1 | 6 - 7 | 1979 | 62 | |
| XM_008983196.2 | XP_008981444.1 | 6 - 7 | 1909 | 62 | ||
More Information
Set Membership: |
|

LOC100391167
Genomic Map