Assay Details
| Gene Symbol: ALKAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758893.2 | XP_002758939.1 | 3 - 4 | 521 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALKAL1| Gene Symbol: ALKAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758893.2 | XP_002758939.1 | 3 - 4 | 521 | 97 | |