Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970187.1 | XP_017825676.1 | 8 - 9 | 1590 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGG| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970187.1 | XP_017825676.1 | 8 - 9 | 1590 | 95 | |
Set Membership: |
|