Assay Details
| Gene Symbol: PPARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743854.3 | XP_002743900.1 | 5 - 6 | 821 | 79 | |
| XM_008980235.2 | XP_008978483.1 | 6 - 7 | 1051 | 79 | ||
| XM_008980242.2 | XP_008978490.1 | 5 - 6 | 1000 | 79 | ||
| XM_017964191.1 | XP_017819680.1 | 3 - 4 | 453 | 79 | ||
More Information
Set Membership: |
|

PPARA
Genomic Map