Assay Details
| Gene Symbol: TICAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204343.1 | NP_001191272.1 | 1 | 140 | 81 | |
| XM_008991631.2 | XP_008989879.1 | 2 | 1694 | 81 | ||
More Information
Set Membership: |
|

TICAM2
Genomic Map