Assay Details
| Gene Symbol: LOC100410987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756516.3 | XP_002756562.1 | 6 - 7 | 1849 | 56 | |
| XM_017979229.1 | XP_017834718.1 | 7 - 8 | 952 | 56 | ||
More Information
Set Membership: |
|

LOC100410987
Genomic Map