Assay Details
| Gene Symbol: LOC100410987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756516.3 | XP_002756562.1 | 8 - 9 | 2114 | 70 | |
| XM_017979229.1 | XP_017834718.1 | 9 - 10 | 1217 | 70 | ||
More Information
Set Membership: |
|

LOC100410987
Genomic Map