Assay Details
| Gene Symbol: DDAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746353.4 | XP_002746399.1 | 6 - 7 | 1025 | 69 | |
| XM_008994169.2 | XP_008992417.1 | 6 - 7 | 922 | 69 | ||
| XM_008994170.2 | XP_008992418.1 | 6 - 7 | 1018 | 69 | ||
More Information
Set Membership: |
|

DDAH2
Genomic Map