Assay Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733360.2 | XP_003733408.1 | 1 - 2 | 192 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003733360.2 transcripts.
Set Membership: |
|

C1QA
Genomic Map