Assay Details
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969799.1 | XP_017825288.1 | 1 - 2 | 345 | 68 | |
| XM_017969800.1 | XP_017825289.1 | 1 - 2 | 359 | 68 | ||
More Information
Set Membership: |
|

CCNH
Genomic Map