Assay Details
| Gene Symbol: NACAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751399.3 | XP_002751445.1 | 3 - 4 | 4124 | 62 | |
| XM_009002145.2 | XP_009000393.1 | 2 - 3 | 6553 | 62 | ||
More Information
Set Membership: |
|

NACAD
Genomic Map