Assay Details
| Gene Symbol: GBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001646.2 | XP_008999894.1 | 10 - 11 | 1887 | 85 | |
| XM_017974906.1 | XP_017830395.1 | 11 - 12 | 2025 | 85 | ||
More Information
Set Membership: |
|

GBP5
Genomic Map