Assay Details
| Gene Symbol: NOCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745404.3 | XP_002745450.1 | 2 - 3 | 840 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOCT| Gene Symbol: NOCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745404.3 | XP_002745450.1 | 2 - 3 | 840 | 94 | |
Set Membership: |
|