Assay Details
| Gene Symbol: B3GLCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973022.1 | XP_017828511.1 | 14 - 15 | 1313 | 60 | |
| XM_017973023.1 | XP_017828512.1 | 14 - 15 | 1236 | 60 | ||
| XM_017973024.1 | XP_017828513.1 | 13 - 14 | 1193 | 60 | ||
More Information
Set Membership: |
|

B3GLCT
Genomic Map