Assay Details
| Gene Symbol: CADM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760154.2 | XP_002760200.1 | 9 - 10 | 1347 | 65 | |
| XM_002760155.3 | XP_002760201.1 | 8 - 9 | 1268 | 65 | ||
More Information
Set Membership: |
|

CADM3
Genomic Map