Assay Details
| Gene Symbol: CCL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749863.3 | XP_002749909.1 | 3 - 4 | 334 | 76 | |
| XM_008999640.1 | XP_008997888.1 | 3 - 4 | 331 | 76 | ||
More Information
Set Membership: |
|

CCL20
Genomic Map