Assay Details
| Gene Symbol: METTL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755153.3 | XP_002755199.1 | 3 - 4 | 798 | 83 | |
| XM_009007859.2 | XP_009006107.1 | 3 - 4 | 609 | 83 | ||
| XM_009007860.2 | XP_009006108.1 | 3 - 4 | 626 | 83 | ||
| XM_017977971.1 | XP_017833460.1 | 3 - 4 | 720 | 83 | ||
More Information
Set Membership: |
|

METTL15
Genomic Map