Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749669.3 | XP_002749715.1 | 3 - 4 | 716 | 106 | |
| XM_002749670.3 | XP_002749716.1 | 3 - 4 | 2480 | 106 | ||
| GenBank mRNA | GQ284838.1 | - | 3 - 4 | 583 | 106 | |
More Information
Set Membership: |
|

CTLA4
Genomic Map