Assay Details
| Gene Symbol: ARL4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972375.1 | XP_017827864.1 | 3 | 1979 | 96 | |
| GenBank mRNA | EF215087.1 | - | 1 | 522 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARL4D| Gene Symbol: ARL4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972375.1 | XP_017827864.1 | 3 | 1979 | 96 | |
| GenBank mRNA | EF215087.1 | - | 1 | 522 | 96 | |
Set Membership: |
|