Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759086.3 | XP_002759132.1 | 2 - 3 | 333 | 102 | |
| XM_008982922.1 | XP_008981170.1 | 2 - 3 | 522 | 102 | ||
More Information
Set Membership: |
|

TP53INP1
Genomic Map