Assay Details
| Gene Symbol: NT5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732703.3 | XP_003732751.1 | 6 - 7 | 1404 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NT5E| Gene Symbol: NT5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732703.3 | XP_003732751.1 | 6 - 7 | 1404 | 64 | |
Set Membership: |
|