Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994990.1 | XP_008993238.1 | 2 - 3 | 360 | 89 | |
| XM_008994991.1 | XP_008993239.1 | 2 - 3 | 360 | 89 | ||
| XM_017971371.1 | XP_017826860.1 | 2 - 3 | 200 | 89 | ||
More Information
Set Membership: |
|

HDDC2
Genomic Map