Assay Details
| Gene Symbol: MRPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759024.4 | XP_002759070.2 | 2 - 3 | 410 | 117 | |
| XM_008982821.2 | XP_008981069.2 | 2 - 3 | 422 | 117 | ||
| XM_008982822.2 | XP_008981070.2 | 2 - 3 | 363 | 117 | ||
| XM_008982823.2 | XP_008981071.2 | 2 - 3 | 348 | 117 | ||
More Information
Set Membership: |
|

MRPS28
Genomic Map