Assay Details
| Gene Symbol: RASSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760730.2 | XP_002760776.1 | 3 - 4 | 790 | 77 | |
| XM_008985565.1 | XP_008983813.1 | 2 - 3 | 685 | 77 | ||
More Information
Set Membership: |
|

RASSF5
Genomic Map