Assay Details
| Gene Symbol: ANGPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759302.3 | XP_002759348.1 | 6 - 7 | 1347 | 72 | |
| XM_008983362.2 | XP_008981610.1 | 6 - 7 | 2803 | 72 | ||
| XM_008983365.2 | XP_008981613.1 | 6 - 7 | 2808 | 72 | ||
| XM_008983366.2 | XP_008981614.1 | 6 - 7 | 3097 | 72 | ||
More Information
Set Membership: |
|

ANGPT1
Genomic Map