Assay Details
| Gene Symbol: C16H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982945.2 | XP_008981193.1 | 2 - 3 | 411 | 75 | |
| XM_017965475.1 | XP_017820964.1 | 2 - 3 | 413 | 75 | ||
More Information
Set Membership: |
|

C16H8orf33
Genomic Map