Assay Details
| Gene Symbol: PMVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760048.4 | XP_002760094.1 | 2 - 3 | 486 | 75 | |
| XM_008984493.2 | XP_008982741.1 | 3 - 4 | 261 | 75 | ||
| XM_017966314.1 | XP_017821803.1 | 2 - 3 | 87 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017966314.1 transcripts.
Set Membership: |
|

PMVK
Genomic Map