Assay Details
| Gene Symbol: MRPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746560.4 | XP_002746606.1 | 2 - 3 | 433 | 73 | |
| XM_008994450.2 | XP_008992698.1 | 2 - 3 | 385 | 73 | ||
| XM_008994451.2 | XP_008992699.1 | 2 - 3 | 248 | 73 | ||
| XM_008994452.2 | XP_008992700.1 | 2 - 3 | 242 | 73 | ||
More Information
Set Membership: |
|

MRPL2
Genomic Map